Learn More
Waters Corp Waters Column, XBridge BEH Amide, 130Å, 2.5 μm, 3 mm x 100 mm
Supplier:  Waters Corp 186006095
Encompass_Preferred
These columns are engineered for highly polar compounds like carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11, for diverse analyte separation. Packed in ultra-low dispersion hardware, these 2.5 μm eXtended Performance columns deliver exceptional HPLC performance and enable smooth UPLC adoption.
  • Retains highly polar compounds such as carbohydrates and sugars.
  • Chemically stable amide phase (pH 2–11) for broad analyte separation.
  • Ultra-low dispersion hardware ensures exceptional performance and throughput for HPLC and UPLC.
  • High-performance reversed-phase LC for amide-containing compounds.
  • Compatible with all major liquid chromatography systems.
  • Single-use format offers convenience and cost-effectiveness.
  • Hybrid silica substrate provides enhanced durability.
Catalog No. 50-179-513