Learn More
Waters Corp WATERS CORPORATION XBridge BEH Amide XP Column, 130Å, 2.5 μm, 2.1 mm x 30 mm
Catalog No. 50179245 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Amide columns are designed to retain extremely polar compounds like carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11. Packed in ultra-low dispersion hardware, these 2.5 μm eXtended Performance (XP) columns offer exceptional separation for HPLC assays and a seamless transition to UPLC adoption.
  • Packed in ultra-low dispersion hardware, offering exceptional separation performance for HPLC assays.
  • Hydrophilic Interaction (HILIC) separation mode with hybrid, spherical particle substrate.
  • Maximum pressure of 18000 psi (1240 Bar).
  • Low silanol activity.
  • Pore size of 130 Å and surface area of 185.
  • Compatible with UPLC and UHPLC systems, featuring BEH particle technology and eCord.
  • USP Classification: L68.
  • 12% carbon load.
Catalog No. 50-179-245 Supplier Waters Corp Supplier No. 176002586
XBridge BEH Amide columns are designed to retain extremely polar compounds like carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11. Packed in ultra-low dispersion hardware, these 2.5 μm eXtended Performance (XP) columns offer exceptional separation for HPLC assays and a seamless transition to UPLC adoption.
  • Packed in ultra-low dispersion hardware, offering exceptional separation performance for HPLC assays.
  • Hydrophilic Interaction (HILIC) separation mode with hybrid, spherical particle substrate.
  • Maximum pressure of 18000 psi (1240 Bar).
  • Low silanol activity.
  • Pore size of 130 Å and surface area of 185.
  • Compatible with UPLC and UHPLC systems, featuring BEH particle technology and eCord.
  • USP Classification: L68.
  • 12% carbon load.