Learn More
Waters Corp Waters Corporation XBridge BEH Amide Method Validation Kit, 130Å, 2.5 μm, 3 pk
Supplier:  Waters Corp 186006231
Encompass_Preferred
This method validation kit consists of three batches of BEH Amide columns packed with three different batches of sorbent.
  • Includes three batches of BEH Amide columns with varying sorbent batches to facilitate comprehensive method validation.
Catalog No. 50-201-586