Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge BEH Amide XP Column, 3.0 x 30 mm, 2.5 µm, 130 Å

Catalog No. 50179249 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XBridge BEH Amide XP 2.5 µm columns enhance transitions between HPLC and UPLC™ technology. Compatible with various LC systems, they improve productivity at intermediate backpressures. These columns provide exceptional sample loading capacity for applications like impurity profiling, metabolite identification, and stability-indicating assays. Methods can be scaled and transferred between HPLC and UPLC columns.

  • Dimensions: 3.0 x 30 mm
  • Particle size: 2.5 µm
  • Pore size: 130 Å
  • Ligand type: Trifunctional Carbamoyl
  • Ligand density: 7.5 µmol/m2
  • Carbon load: 12%
  • pH range: 2 to 11
  • Low pH temperature limit: 90 °C
  • High pH temperature limit: 90 °C
  • Surface area: 185 m2/g

Catalog No. 50-179-249 Supplier Waters Corp Supplier No. 176002590
May include imposed supplier surcharges.
Add to Cart
Add to Cart

XBridge BEH Amide XP 2.5 µm columns enhance transitions between HPLC and UPLC™ technology. Compatible with various LC systems, they improve productivity at intermediate backpressures. These columns provide exceptional sample loading capacity for applications like impurity profiling, metabolite identification, and stability-indicating assays. Methods can be scaled and transferred between HPLC and UPLC columns.

  • Dimensions: 3.0 x 30 mm
  • Particle size: 2.5 µm
  • Pore size: 130 Å
  • Ligand type: Trifunctional Carbamoyl
  • Ligand density: 7.5 µmol/m2
  • Carbon load: 12%
  • pH range: 2 to 11
  • Low pH temperature limit: 90 °C
  • High pH temperature limit: 90 °C
  • Surface area: 185 m2/g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.