Learn More
Waters Corp Waters XBridge BEH Amide Column, 130Å, 2.5 μm, 3 mm X 50 mm
Supplier:  Waters Corp 186006093
Encompass_Preferred
These columns retain extremely polar compounds, including carbohydrates and sugars, utilizing a chemically stable, trifunctionally bonded amide phase stable across a pH range of 2 to 11. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance (XP) columns deliver exceptional separation performance and robustness for HPLC assays, facilitating a seamless transition to UPLC.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Features a chemically stable amide phase, stable from pH 2 to 11.
  • Packed in ultra-low dispersion hardware for exceptional separation performance, robustness, and throughput.
  • Built on BEH particle technology for increased hydrolytic stability and particle strength, minimizing LC/MS bleed.
Catalog No. 50-179-511