Learn More
Waters Corp Waters Column, Amide, 130Å, 2.5 μm, 4.6 mm x 75 mm
Catalog No. 50201536 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XBridge BEH Amide columns are engineered for the retention of highly polar compounds, including carbohydrates and sugars. Featuring a chemically stable, trifunctionally bonded amide phase, they maintain stability across a pH range of 2 to 11. The 2.5 μm eXtended Performance (XP) design ensures exceptional separation and robustness for HPLC assays, facilitating a seamless transition to UPLC methods.
  • Retains highly polar compounds.
  • Stable amide phase (pH 2-11).
  • Ultra-low dispersion hardware.
  • Exceptional HPLC assay performance.
  • Smooth transition to UPLC.
  • Rugged, repeatable HPLC/UHPLC performance.
  • BEH particle technology.
  • Supports reversed-phase and HILIC.
Catalog No. 50-201-536 Supplier Waters Corp Supplier No. 186006098
These XBridge BEH Amide columns are engineered for the retention of highly polar compounds, including carbohydrates and sugars. Featuring a chemically stable, trifunctionally bonded amide phase, they maintain stability across a pH range of 2 to 11. The 2.5 μm eXtended Performance (XP) design ensures exceptional separation and robustness for HPLC assays, facilitating a seamless transition to UPLC methods.
  • Retains highly polar compounds.
  • Stable amide phase (pH 2-11).
  • Ultra-low dispersion hardware.
  • Exceptional HPLC assay performance.
  • Smooth transition to UPLC.
  • Rugged, repeatable HPLC/UHPLC performance.
  • BEH particle technology.
  • Supports reversed-phase and HILIC.