Learn More
Waters Corp Waters Column, Amide, 130Å, 2.5 μm, 2.1 mm × 100 mm
Catalog No. 50179509 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns retain extremely polar compounds, including carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11, enabling separation of polar analytes. The 2.5 μm eXtended Performance (XP) columns use ultra-low dispersion hardware for exceptional separation performance, robustness, and seamless UPLC transition.
  • Facilitates easy and convenient method transfer between HPLC and UPLC.
  • Withstands high pressures up to 18,000 psi (1240 bar).
  • Minimizes retention loss and peak shape degradation compared to conventional columns.
  • Ensures exceptional column lifetime through proprietary bonding and end-capping procedures.
  • Provides ligand stability and chromatographic reproducibility at both low and high pH conditions.
  • Delivers consistent and reproducible performance throughout its lifespan.
Catalog No. 50-179-509 Supplier Waters Corp Supplier No. 186006091
These columns retain extremely polar compounds, including carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11, enabling separation of polar analytes. The 2.5 μm eXtended Performance (XP) columns use ultra-low dispersion hardware for exceptional separation performance, robustness, and seamless UPLC transition.
  • Facilitates easy and convenient method transfer between HPLC and UPLC.
  • Withstands high pressures up to 18,000 psi (1240 bar).
  • Minimizes retention loss and peak shape degradation compared to conventional columns.
  • Ensures exceptional column lifetime through proprietary bonding and end-capping procedures.
  • Provides ligand stability and chromatographic reproducibility at both low and high pH conditions.
  • Delivers consistent and reproducible performance throughout its lifespan.