Learn More
Waters Corp XBridge BEH Amide Method Validation Kit, 130Å, 2.5 µm, 2.1 mm X 100 mm, 3/pk
Catalog No. 50179597 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three batches of XBridge BEH Amide columns packed with three different batches of sorbent
Catalog No. 50-179-597 Supplier Waters Corp Supplier No. 186006228
This method validation kit consists of three batches of XBridge BEH Amide columns packed with three different batches of sorbent