Learn More
Waters Corp Column, Amide, 2.5 μm, 130 Å Pore, 4.6 mm x 50 mm
Supplier:  Waters Corp 186006097
Encompass_Preferred
These BEH Amide columns are designed for the retention of extremely polar compounds like carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11, allowing separation of polar analytes across a wide range of polarity. Packed in ultra-low dispersion hardware, the 2.5 μm XP columns provide exceptional separation performance and robustness for HPLC assays, with a seamless path to UPLC adoption.
  • Designed to retain extremely polar compounds.
  • Chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11.
  • Separates polar analytes across a wide range of polarity.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance (XP) for exceptional separation and robustness.
  • Seamless transition path toward UPLC adoption.
Catalog No. 50-201-535