Learn More
Waters Corp Waters XBridge BEH Amide Column, 130Å, 2.5 μm, 2.1 x 75 mm
Catalog No. 50179508 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars. Featuring a chemically stable, trifunctionally bonded amide phase, they are stable across a pH range of 2 to 11, allowing separation of diverse polar analytes. These 2.5 μm XP columns are packed in ultra-low dispersion hardware, providing excellent separation, robustness, and throughput for HPLC assays, while offering an upgrade path to UPLC.
  • Retains extremely polar compounds like carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable across a wide pH range (pH 2 to 11).
  • Enables separation of diverse polar analytes by polarity, structure, and pKa.
  • Packed in ultra-low dispersion hardware for eXtended performance (XP).
  • Delivers exceptional separation performance, robustness, and throughput for HPLC assays.
Catalog No. 50-179-508 Supplier Waters Corp Supplier No. 186006090
BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars. Featuring a chemically stable, trifunctionally bonded amide phase, they are stable across a pH range of 2 to 11, allowing separation of diverse polar analytes. These 2.5 μm XP columns are packed in ultra-low dispersion hardware, providing excellent separation, robustness, and throughput for HPLC assays, while offering an upgrade path to UPLC.
  • Retains extremely polar compounds like carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable across a wide pH range (pH 2 to 11).
  • Enables separation of diverse polar analytes by polarity, structure, and pKa.
  • Packed in ultra-low dispersion hardware for eXtended performance (XP).
  • Delivers exceptional separation performance, robustness, and throughput for HPLC assays.