Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp HPLC Column, XBridge BEH Amide, 130 Å, 2.5 μm, 3 mm x 100 mm, 3/pk

Catalog No. 50179252 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XBridge BEH Amide columns retain extremely polar compounds like carbohydrates and sugars. They feature a chemically stable amide phase, stable from pH 2 to 11, allowing separation of polar analytes across a broad range of polarity and pKa. Packed in ultra-low dispersion hardware, 2.5 μm XP columns provide exceptional separation, robustness, and throughput for HPLC assays, enabling a seamless transition to UPLC.

  • Retains polar compounds (carbohydrates, sugars).
  • Chemically stable amide phase.
  • pH stability: 2 to 11.
  • Ultra-low dispersion hardware.
  • Particle size: 2.5 μm (eXtended Performance).
  • Exceptional separation, robustness, and throughput.
  • Seamless transition to UPLC.
  • Carbon load: 12.0%.
  • Inner diameter: 3 mm.
  • Length: 100 mm.
  • Pore size: 130 Å.
  • USP number: L1.
  • Packaged as 3 units.

Catalog No. 50-179-252 Supplier Waters Corp Supplier No. 176002593
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XBridge BEH Amide columns retain extremely polar compounds like carbohydrates and sugars. They feature a chemically stable amide phase, stable from pH 2 to 11, allowing separation of polar analytes across a broad range of polarity and pKa. Packed in ultra-low dispersion hardware, 2.5 μm XP columns provide exceptional separation, robustness, and throughput for HPLC assays, enabling a seamless transition to UPLC.

  • Retains polar compounds (carbohydrates, sugars).
  • Chemically stable amide phase.
  • pH stability: 2 to 11.
  • Ultra-low dispersion hardware.
  • Particle size: 2.5 μm (eXtended Performance).
  • Exceptional separation, robustness, and throughput.
  • Seamless transition to UPLC.
  • Carbon load: 12.0%.
  • Inner diameter: 3 mm.
  • Length: 100 mm.
  • Pore size: 130 Å.
  • USP number: L1.
  • Packaged as 3 units.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.