Learn More
Waters Corp Waters XBridge BEH Amide Column, 130Å, 2.5 μm, 2.1 x 50 mm
Supplier:  Waters Corp 186006089
Encompass_Preferred
This column is designed to retain extremely polar compounds like carbohydrates and sugars, utilizing a chemically stable, trifunctionally bonded amide phase with pH stability from 2 to 11. Its 2.5 μm eXtended Performance (XP) particles deliver exceptional separation for HPLC assays and ease transition to UPLC, enabling high resolution and speed in complex sample analysis.
  • Retains extremely polar compounds (e.g., carbohydrates and sugars).
  • Chemically stable amide phase, stable from pH 2 to 11.
  • eXtended Performance (XP) particles for exceptional HPLC separation.
  • Seamless transition to UPLC due to consistent selectivity.
  • High-resolution, high-speed analysis of carbohydrates with 2.5 μm particle and 130Å pore size.
  • Utilizes ethylene bridged hybrid (BEH) particle technology for HILIC stability and versatility.
Catalog No. 50-179-507