Learn More
Waters Corp Waters Column, C18, 130Å, 2.5 μm, 2.1 mm, 150 mm
Catalog No. 50466417 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent enables HPLC method development across a mobile phase pH range of 1 to 12. With superior low pH stability and ultra-low column bleed, its ultra-low dispersion hardware ensures exceptional separation performance and robustness for HPLC assays, facilitating seamless transition toward UPLC adoption.
  • Trifunctionally bonded C18 ligand for universal separation.
  • Usable pH range of 1 to 12.
  • Superior low pH stability; ultra-low column bleed.
  • Ultra-low dispersion hardware ensures exceptional performance.
  • Withstands pressures up to 18,000 psi (1240 Bar).
  • Ideal for QC methods or LC/MS assay development.
  • Improved pH stability extends column lifetimes.
  • Maximizes particle efficiency for excellent peak shape.
  • Proprietary bonding ensures ligand stability and reproducibility at low pH.
Catalog No. 50-466-417 Supplier Waters Corp Supplier No. 186006709
The XBridge BEH C18 sorbent enables HPLC method development across a mobile phase pH range of 1 to 12. With superior low pH stability and ultra-low column bleed, its ultra-low dispersion hardware ensures exceptional separation performance and robustness for HPLC assays, facilitating seamless transition toward UPLC adoption.
  • Trifunctionally bonded C18 ligand for universal separation.
  • Usable pH range of 1 to 12.
  • Superior low pH stability; ultra-low column bleed.
  • Ultra-low dispersion hardware ensures exceptional performance.
  • Withstands pressures up to 18,000 psi (1240 Bar).
  • Ideal for QC methods or LC/MS assay development.
  • Improved pH stability extends column lifetimes.
  • Maximizes particle efficiency for excellent peak shape.
  • Proprietary bonding ensures ligand stability and reproducibility at low pH.