Learn More
Waters Corp Waters XBridge BEH C18 Method Validation Kit, 2.5 μm, 2.1 mm x 150 mm, 3 pk
Supplier:  Waters Corp 186006757
Encompass_Preferred
This method validation kit consists of three XBridge BEH C18 columns packed with three different batches of sorbent, suitable for HPLC method development. It offers a universal solution for most HPLC separations, providing a wide usable pH range and superior stability.
  • Suitable for HPLC method development
  • Can be used over a mobile phase pH range of 1 to 12
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations
  • Offers the widest usable pH range, superior low pH stability, and ultra-low column bleed
  • Packed in ultra-low dispersion hardware
  • eXtended Performance (XP) columns deliver exceptional separation performance, robustness, and throughput for HPLC assays
  • Provides a seamless transition path toward UPLC adoption
Catalog No. 50-466-459