Learn More
Waters Corp XBridge BEH C18 Method Validation Kit, 130Å, 2.5 µm, 3 mm X 150 mm, 3/pk
Catalog No. 50466460 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three Xbridge BEH C18 columns packed with three different batches of sorbent. As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-460 Supplier Waters Corp Supplier No. 186006758
This method validation kit consists of three Xbridge BEH C18 columns packed with three different batches of sorbent. As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.