Learn More
Waters Corp Waters XBridge BEH C18 Column, 2.5 μm, 3 mm x 150 mm
Supplier:  Waters Corp 186006710
Encompass_Preferred
The XBridge BEH C18 sorbent is for HPLC method development, usable across a mobile phase pH range of 1–12. Its trifunctionally bonded C18 ligand provides a universal column for most HPLC separations, offering wide pH range, superior low pH stability, and ultra–low column bleed.
  • Offers superior pH stability over a wide range (0–12) with high efficiency.
  • Provides unmatched column lifetime and flexibility across varied mobile phase pH and temperatures.
  • Maximizes productivity for quality control or developing LC/MS assays.
  • Delivers excellent peak shape and capacity by maximizing particle efficiency.
  • Minimizes retention loss and degradation with proprietary bonding, ensuring ligand stability and reproducibility at low pH.
Catalog No. 50-466-418