Learn More
Waters Corp Waters Column, C18, 2.5 μm, 3 mm, 150 mm
Catalog No. 50465920 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent, with its trifunctionally bonded C18 ligand, is designed for HPLC method development across a pH range of 1-12. It provides superior low pH stability and ultra-low column bleed. These 2.5 μm XP columns, in ultra-low dispersion hardware, deliver exceptional separation performance, robustness, and throughput for HPLC, aiding UPLC transition.
  • Optimized for method development via BEH technology.
  • Hybrid particles reduce silanol concentration and lower USP peak tailing for bases.
  • Internal bridging groups provide chemical and mechanical stability.
  • Hydrophobicity increases stability in high pH ranges.
  • Virtually eliminates LC/MS bleed; enhanced hydrolytic stability and particle strength.
  • Withstands up to 18000 psi.
  • Long column lifetime reduces replacement needs.
  • 18% carbon load.
Catalog No. 50-465-920 Supplier Waters Corp Supplier No. 176002880
The XBridge BEH C18 sorbent, with its trifunctionally bonded C18 ligand, is designed for HPLC method development across a pH range of 1-12. It provides superior low pH stability and ultra-low column bleed. These 2.5 μm XP columns, in ultra-low dispersion hardware, deliver exceptional separation performance, robustness, and throughput for HPLC, aiding UPLC transition.
  • Optimized for method development via BEH technology.
  • Hybrid particles reduce silanol concentration and lower USP peak tailing for bases.
  • Internal bridging groups provide chemical and mechanical stability.
  • Hydrophobicity increases stability in high pH ranges.
  • Virtually eliminates LC/MS bleed; enhanced hydrolytic stability and particle strength.
  • Withstands up to 18000 psi.
  • Long column lifetime reduces replacement needs.
  • 18% carbon load.