Learn More
Waters Corp Waters Column, C18, 130 Å, 2.5 μm, 4.6 mm × 150 mm
Catalog No. 50466419 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for HPLC method development, offering universal C18 separation across a mobile phase pH range of 1-12. It provides superior low pH stability, ultra-low column bleed, and is packed in ultra-low dispersion hardware for exceptional performance and throughput in HPLC assays, facilitating transition to UPLC.
  • Offers universal C18 separation for HPLC method development across a mobile phase pH range of 1-12.
  • Provides superior low pH stability and ultra-low column bleed for enhanced performance.
  • Packed in ultra-low dispersion hardware for exceptional separation performance and robustness.
  • Delivers reproducible results, maximizing particle efficiency for unmatched peak shape and increased column lifetime.
Catalog No. 50-466-419 Supplier Waters Corp Supplier No. 186006711
This column is designed for HPLC method development, offering universal C18 separation across a mobile phase pH range of 1-12. It provides superior low pH stability, ultra-low column bleed, and is packed in ultra-low dispersion hardware for exceptional performance and throughput in HPLC assays, facilitating transition to UPLC.
  • Offers universal C18 separation for HPLC method development across a mobile phase pH range of 1-12.
  • Provides superior low pH stability and ultra-low column bleed for enhanced performance.
  • Packed in ultra-low dispersion hardware for exceptional separation performance and robustness.
  • Delivers reproducible results, maximizing particle efficiency for unmatched peak shape and increased column lifetime.