Learn More
Waters Corp XBridge BEH C18 Column, 130 Å, 3.5 μm, 2.1 mm X 150 mm
Supplier:  Waters Corp 186003023RF
Encompass_Preferred
The XBridge BEH C18 Column is a reliable workhorse for HPLC method development, offering broad mobile phase pH compatibility (1–12). Its trifunctionally bonded C18 ligand provides a universal column for most HPLC separations with superior low pH stability and ultra-low column bleed. This column includes an electronic eConnect column tag, an NFC-enabled device for automatic identification, tracking, and digital tracing of usage history, maximizing benefits with Waters Alliance iS System and Empower software.
  • Universal column for most HPLC separations
  • Widest usable pH range (1–12)
  • Superior low pH stability and ultra-low column bleed
  • Features an eConnect column tag for automatic identification and usage history tracking
  • NFC-enabled for digital tracing
  • Trifunctionally bonded C18 ligand
  • BEH Technology
  • Hybrid particle substrate
Catalog No. 50-239-0751