Learn More
Waters Corp Waters Corporation XBridge BEH C18 Column, 130 Å, 5 µm, 4.6 mm x 100 mm
Catalog No. 502392219 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge BEH C18 Column is designed for high-performance liquid chromatography (HPLC) applications, featuring a 130 Å pore size, 5 µm particle size, and 4.6 mm x 100 mm dimensions. It incorporates eConnect technology, a second-generation smart-column system that uses an electronic tag on the column body to store product-specific information and facilitate data management.
  • Ready for use direct from the box.
  • eConnect tag carries all column-specific identifying information, simplifying setup and reducing manual data entry.
  • Permanent, tamper-proof tag, locked onto the column body.
  • NFC-capable tag provides convenient access to product-support information.
  • Designed for compliant laboratories and assists in migrating towards an all-digital platform.
  • Automatically identifies, verifies, and traces column usage history (together with Alliance iS and Empower CDS).
Catalog No. 50-239-2219 Supplier Waters Corp Supplier No. 186003115RF
This XBridge BEH C18 Column is designed for high-performance liquid chromatography (HPLC) applications, featuring a 130 Å pore size, 5 µm particle size, and 4.6 mm x 100 mm dimensions. It incorporates eConnect technology, a second-generation smart-column system that uses an electronic tag on the column body to store product-specific information and facilitate data management.
  • Ready for use direct from the box.
  • eConnect tag carries all column-specific identifying information, simplifying setup and reducing manual data entry.
  • Permanent, tamper-proof tag, locked onto the column body.
  • NFC-capable tag provides convenient access to product-support information.
  • Designed for compliant laboratories and assists in migrating towards an all-digital platform.
  • Automatically identifies, verifies, and traces column usage history (together with Alliance iS and Empower CDS).