Learn More
Waters Corp XBridge BEH C18 Column, 130 Å, 3.5 μm, 4.6 mm × 250 mm
Catalog No. 502391896 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This versatile HPLC column is designed for method development, offering a wide mobile phase pH range of 1–12. Its trifunctionally bonded C18 ligand ensures superior low pH stability and ultra-low column bleed, making it suitable for most HPLC separations. An electronic eConnect column tag allows for automatic identification, tracking, and digital tracing of use history, enhancing compatibility with any HPLC system, especially when paired with the Alliance iS System running Empower software.
  • HPLC method development workhorse with trifunctionally bonded C18 ligand.
  • Wide usable pH range (1–12) with superior low pH stability.
  • Features ultra-low column bleed.
  • eConnect column tag for automatic identification, tracking, and digital tracing.
  • Compatible with any HPLC system; enhanced with compatible software.
Catalog No. 50-239-1896 Supplier Waters Corp Supplier No. 186003943RF
Add to Cart
Add to Cart
This versatile HPLC column is designed for method development, offering a wide mobile phase pH range of 1–12. Its trifunctionally bonded C18 ligand ensures superior low pH stability and ultra-low column bleed, making it suitable for most HPLC separations. An electronic eConnect column tag allows for automatic identification, tracking, and digital tracing of use history, enhancing compatibility with any HPLC system, especially when paired with the Alliance iS System running Empower software.
  • HPLC method development workhorse with trifunctionally bonded C18 ligand.
  • Wide usable pH range (1–12) with superior low pH stability.
  • Features ultra-low column bleed.
  • eConnect column tag for automatic identification, tracking, and digital tracing.
  • Compatible with any HPLC system; enhanced with compatible software.