Learn More
Waters Corp XBridge Peptide BEH C18 XP Column, 300 Å, 2.5 μm
Catalog No. 502038903 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These BEH C18 columns offer a wide pH range (1–12) for diverse compound separations. Their trifunctional bonding and end-capping ensure stability, reproducibility, and efficiency across various chromatographic conditions. They provide excellent batch-to-batch reproducibility, essential for reliable and robust analytical methods.
  • Wide pH range (1–12) for diverse compound analysis.
  • Trifunctional bonding and end-capping for enhanced stability and efficiency.
  • Excellent batch-to-batch reproducibility.
  • Consistent performance across manufacturing lots.
  • Ideal for robust method development.
  • Suitable for analyzing various compounds, including rosuvastatin and impurities.
Catalog No. 50-203-8903 Supplier Waters Corp Supplier No. 186009075
These BEH C18 columns offer a wide pH range (1–12) for diverse compound separations. Their trifunctional bonding and end-capping ensure stability, reproducibility, and efficiency across various chromatographic conditions. They provide excellent batch-to-batch reproducibility, essential for reliable and robust analytical methods.
  • Wide pH range (1–12) for diverse compound analysis.
  • Trifunctional bonding and end-capping for enhanced stability and efficiency.
  • Excellent batch-to-batch reproducibility.
  • Consistent performance across manufacturing lots.
  • Ideal for robust method development.
  • Suitable for analyzing various compounds, including rosuvastatin and impurities.