Learn More
Waters Corp XBridge Preparative Column, BEH C18, 10 x 150 mm, 10 μm
Catalog No. 502390891 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These OBD Preparative Columns feature uniform, optimum packed bed density, enabling direct scalability from analytical to preparative chromatography. They offer excellent column lifetime and consistent performance, resulting in superior resolution, peak shape, and mass loading.
  • Uniform packed bed density.
  • Scalable from analytical to preparative chromatography.
  • Excellent column lifetime and consistent performance.
  • Delivers superior resolution, peak shape, and mass loading.
  • BEH Technology™ for broad compound compatibility.
  • Exceptional peak shape for basic analytes at high pH.
  • Wide pH stability (1 to 12).
  • High temperature stability (up to 80 °C low pH, 60 °C high pH).
Catalog No. 50-239-0891 Supplier Waters Corp Supplier No. 186008210
These OBD Preparative Columns feature uniform, optimum packed bed density, enabling direct scalability from analytical to preparative chromatography. They offer excellent column lifetime and consistent performance, resulting in superior resolution, peak shape, and mass loading.
  • Uniform packed bed density.
  • Scalable from analytical to preparative chromatography.
  • Excellent column lifetime and consistent performance.
  • Delivers superior resolution, peak shape, and mass loading.
  • BEH Technology™ for broad compound compatibility.
  • Exceptional peak shape for basic analytes at high pH.
  • Wide pH stability (1 to 12).
  • High temperature stability (up to 80 °C low pH, 60 °C high pH).