Learn More
Waters Corp Waters OBD Prep Column, BEH C18, 5 μm, 10 mm X 150 mm, 130 Å
Supplier:  Waters Corp 186008166
Encompass_Preferred
This OBD Prep Column is designed for HPLC method development, offering superior low pH stability and ultra-low column bleed across a mobile phase pH range of 1-12. It enables efficient lab-scale separations and immediate scaling from UPLC or HPLC screening to purification.
  • C18 sorbent for HPLC method development, pH 1-12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Enables efficient lab-scale separations and high throughput.
  • Allows immediate scaling from screening to purification.
  • Increases recoveries and productivity, extends column life.
  • Patented packing ensures homogeneous density; no cavities.
  • Resistant to mechanical bed failure and pressure shocks.
  • OBD design for integrity and stability in high-purity isolations.
  • Consistent column performance from stringent production standards.
Catalog No. 50-203-8376