Learn More
Waters Corp Waters OBD Prep Column, BEH C18, 5 μm, 10 mm X 250 mm
Catalog No. 502038377 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent is a workhorse for HPLC method development, usable across a mobile phase pH range of 1–12. Its trifunctionally bonded C18 ligand provides a universal column for most HPLC separations, offering superior low pH stability and ultra-low column bleed. This column facilitates lab-scale separations for increased throughput.
  • Exceptional resistance to mechanical chromatographic bed failure.
  • OBD design resists poor solubility and pressure shocks.
  • Ensures predictable, uniform density profiles and eliminates voids.
  • Delivers consistent column-to-column performance, extending lifetimes and reducing costs.
  • Enables direct scaling from UPLC or HPLC screening to lab-scale purification.
  • Offers higher recoveries while maximizing productivity.
Catalog No. 50-203-8377 Supplier Waters Corp Supplier No. 186008167
The XBridge BEH C18 sorbent is a workhorse for HPLC method development, usable across a mobile phase pH range of 1–12. Its trifunctionally bonded C18 ligand provides a universal column for most HPLC separations, offering superior low pH stability and ultra-low column bleed. This column facilitates lab-scale separations for increased throughput.
  • Exceptional resistance to mechanical chromatographic bed failure.
  • OBD design resists poor solubility and pressure shocks.
  • Ensures predictable, uniform density profiles and eliminates voids.
  • Delivers consistent column-to-column performance, extending lifetimes and reducing costs.
  • Enables direct scaling from UPLC or HPLC screening to lab-scale purification.
  • Offers higher recoveries while maximizing productivity.