Learn More
Waters Corp XBridge VanGuard Cartridge, C18, 130 Å, 3.5 μm, 3.9 × 5 mm
Catalog No. NC1663035 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard Cartridge extends analytical column lifetime and performance by removing particulate contamination. Optimized for 4.6 mm and 3.9 mm I.D. columns with 3.5 μm sorbent, it supports reversed-phase LC. It attaches directly to UPLC Columns, featuring BEH Technology C18, 130Å pore, and 3.5μm particles. Each batch is QC tested for consistent performance.
  • Extends column lifetime by removing particulate contamination.
  • Optimized for 4.6 mm and 3.9 mm I.D. columns with 3.5 μm sorbent.
  • Enhances analytical performance by filtering impurities.
  • Easy to install and use; direct attachment to UPLC columns.
  • Tolerates pressures up to 18,000 psi (1240 bar).
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Consistent performance via peptide map QC testing.
  • Utilizes patented BEH Technology C18.
Catalog No. NC1663035 Supplier Waters Corp Supplier No. 186007768
This VanGuard Cartridge extends analytical column lifetime and performance by removing particulate contamination. Optimized for 4.6 mm and 3.9 mm I.D. columns with 3.5 μm sorbent, it supports reversed-phase LC. It attaches directly to UPLC Columns, featuring BEH Technology C18, 130Å pore, and 3.5μm particles. Each batch is QC tested for consistent performance.
  • Extends column lifetime by removing particulate contamination.
  • Optimized for 4.6 mm and 3.9 mm I.D. columns with 3.5 μm sorbent.
  • Enhances analytical performance by filtering impurities.
  • Easy to install and use; direct attachment to UPLC columns.
  • Tolerates pressures up to 18,000 psi (1240 bar).
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Consistent performance via peptide map QC testing.
  • Utilizes patented BEH Technology C18.