Learn More
Waters Corp Waters XBridge BEH C18 VanGuard Cartridge, 130Å, 3.5 μm
Catalog No. NC1705086 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH C18 VanGuard Cartridges extend the lifetime and enhance the performance of analytical columns by efficiently removing particulate contamination. They are optimized for the protection of 2.1 mm and 3.0 mm I.D. analytical columns utilizing 3.5 μm sorbent particles. These cartridges offer robust method development and exceptional stability across a broad pH range.
  • Efficiently removes particulate contamination from mobile phase stream.
  • Optimized for protecting 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles.
  • Incorporates BEH particle Technology for flexible scaling across chromatography platforms.
  • Ensures robust method development and exceptional stability across a broad pH range of 1 to 12.
  • Manufactured under strict cGMP and ISO 9002 conditions for reliable analysis results.
  • Withstands pressures up to 18,000 psi (1240 Bar).
Catalog No. NC1705086 Supplier Waters Corp Supplier No. 186007766
XBridge BEH C18 VanGuard Cartridges extend the lifetime and enhance the performance of analytical columns by efficiently removing particulate contamination. They are optimized for the protection of 2.1 mm and 3.0 mm I.D. analytical columns utilizing 3.5 μm sorbent particles. These cartridges offer robust method development and exceptional stability across a broad pH range.
  • Efficiently removes particulate contamination from mobile phase stream.
  • Optimized for protecting 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles.
  • Incorporates BEH particle Technology for flexible scaling across chromatography platforms.
  • Ensures robust method development and exceptional stability across a broad pH range of 1 to 12.
  • Manufactured under strict cGMP and ISO 9002 conditions for reliable analysis results.
  • Withstands pressures up to 18,000 psi (1240 Bar).