Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Peptide BEH C18 VanGuard Cartridge, 130Å, 2.5 μm, 3.9 mm × 5 mm

Catalog No. 501528612 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This XBridge Peptide BEH C18 VanGuard Cartridge protects analytical columns. It features a 130Å trifunctionally bonded BEH particle, offering a wide usable pH range, low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.

  • Extends analytical column lifetimes.
  • 130Å trifunctionally bonded BEH particle.
  • Wide usable pH range and low pH stability.
  • Ultra-low column bleed.
  • Suitable for proteomic and peptide mapping.
  • Max temperature: 90°C.
  • Max pressure: 18000 psi (1240 Bar).
  • Particle size: 2.5 μm; Pore size: 130 Å.
  • Compatible with HPLC and UHPLC.
  • Manufactured under cGMP and ISO 9002 guidelines.

Catalog No. 50-152-8612 Supplier Waters Corp Supplier No. 186008989
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This XBridge Peptide BEH C18 VanGuard Cartridge protects analytical columns. It features a 130Å trifunctionally bonded BEH particle, offering a wide usable pH range, low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.

  • Extends analytical column lifetimes.
  • 130Å trifunctionally bonded BEH particle.
  • Wide usable pH range and low pH stability.
  • Ultra-low column bleed.
  • Suitable for proteomic and peptide mapping.
  • Max temperature: 90°C.
  • Max pressure: 18000 psi (1240 Bar).
  • Particle size: 2.5 μm; Pore size: 130 Å.
  • Compatible with HPLC and UHPLC.
  • Manufactured under cGMP and ISO 9002 guidelines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.