Learn More
Waters Corp Waters VanGuard Cartridge, BEH C18, 130 Å, 5 μm, 2.1 mm × 5 mm
Catalog No. 502224195 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard cartridge protects analytical columns in reversed-phase liquid chromatography. It features BEH C18 chemistry, 130 Å pore size, and 5 μm particle size, with dimensions of 2.1 mm × 5 mm. It removes particulate and chemical contaminants from the mobile phase, extending column life and maintaining chromatographic performance.
  • Extends analytical column lifetime and performance by removing particulate and chemical contamination.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns containing 5 μm sorbent.
  • Simple to install and use.
  • Created for UPLC columns in reversed-phase liquid chromatography.
  • Preserves UPLC, UHPLC, and HPLC separation efficiency.
  • Contains 130-pore, 5 μm BEH Technology C18 particles.
Catalog No. 50-222-4195 Supplier Waters Corp Supplier No. 186007769
This VanGuard cartridge protects analytical columns in reversed-phase liquid chromatography. It features BEH C18 chemistry, 130 Å pore size, and 5 μm particle size, with dimensions of 2.1 mm × 5 mm. It removes particulate and chemical contaminants from the mobile phase, extending column life and maintaining chromatographic performance.
  • Extends analytical column lifetime and performance by removing particulate and chemical contamination.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns containing 5 μm sorbent.
  • Simple to install and use.
  • Created for UPLC columns in reversed-phase liquid chromatography.
  • Preserves UPLC, UHPLC, and HPLC separation efficiency.
  • Contains 130-pore, 5 μm BEH Technology C18 particles.