Learn More
Waters Corp Waters XBridge Peptide BEH C18 XP Column, 130 Å, 2.5 μm, 2.1 mm x 50 mm
Catalog No. 501528602 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Peptide BEH C18 XP Column, with 130Å pore size, 2.5 μm particle size, and 2.1 mm x 50 mm dimensions, is optimized and QC tested for peptide separations. It features a trifunctionally bonded BEH particle for wide pH range and low column bleed, ideal for proteomic, peptide mapping, and synthetic peptide applications with UPLC or UHPLC systems. It operates up to 90°C and 1240 Bar.
  • Optimized and QC tested specifically for peptide separations.
  • Features small-pore (130Å) trifunctionally bonded BEH particle for wide pH range and low bleed.
  • Packed in ultra-low dispersion hardware, offering superior separation performance and throughput.
  • Ensures good peak shape, high efficiency, and outstanding stability from cGMP, ISO 9002 accredited production.
  • Each column includes a Performance Test Chromatogram and Certificate of Acceptance.
Catalog No. 50-152-8602 Supplier Waters Corp Supplier No. 186008979
Add to Cart
Add to Cart
This XBridge Peptide BEH C18 XP Column, with 130Å pore size, 2.5 μm particle size, and 2.1 mm x 50 mm dimensions, is optimized and QC tested for peptide separations. It features a trifunctionally bonded BEH particle for wide pH range and low column bleed, ideal for proteomic, peptide mapping, and synthetic peptide applications with UPLC or UHPLC systems. It operates up to 90°C and 1240 Bar.
  • Optimized and QC tested specifically for peptide separations.
  • Features small-pore (130Å) trifunctionally bonded BEH particle for wide pH range and low bleed.
  • Packed in ultra-low dispersion hardware, offering superior separation performance and throughput.
  • Ensures good peak shape, high efficiency, and outstanding stability from cGMP, ISO 9002 accredited production.
  • Each column includes a Performance Test Chromatogram and Certificate of Acceptance.