Learn More
Waters Corp Waters XBridge Peptide BEH C18 XP Column, 130Å, 2.5 μm, 2.1 mm x 150 mm
Catalog No. 501528604 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 XP Column, 130Å, 2.5 μm, 2.1 mm x 150 mm, is optimized and QC tested for peptide separations. It features a small-pore (130Å) trifunctionally bonded BEH particle, which provides the widest usable pH range, superior low pH stability, and ultra-low column bleed. It is designed for proteomic, peptide mapping, and synthetic peptide applications and is recommended for use with low dispersion (UPLC or UHPLC) systems, particularly for LC-MS methods.
  • Optimized for peptide separations, QC tested.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Offers pH range 1 to 12, superior low pH stability, ultra-low column bleed.
  • Exceptional separation performance, robustness, throughput for HPLC assays.
  • Seamless method transfer between HPLC and UPLC.
  • Ultra-low dispersion hardware with 2.5 μm XP particle; withstands 18000 psi max pressure.
Catalog No. 50-152-8604 Supplier Waters Corp Supplier No. 186008981
The XBridge Peptide BEH C18 XP Column, 130Å, 2.5 μm, 2.1 mm x 150 mm, is optimized and QC tested for peptide separations. It features a small-pore (130Å) trifunctionally bonded BEH particle, which provides the widest usable pH range, superior low pH stability, and ultra-low column bleed. It is designed for proteomic, peptide mapping, and synthetic peptide applications and is recommended for use with low dispersion (UPLC or UHPLC) systems, particularly for LC-MS methods.
  • Optimized for peptide separations, QC tested.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Offers pH range 1 to 12, superior low pH stability, ultra-low column bleed.
  • Exceptional separation performance, robustness, throughput for HPLC assays.
  • Seamless method transfer between HPLC and UPLC.
  • Ultra-low dispersion hardware with 2.5 μm XP particle; withstands 18000 psi max pressure.