Learn More
Waters Corp Waters XBridge Peptide BEH C18 XP Column, 130Å, 2.5 μm
Catalog No. 501528605 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 XP Column, 130Å, 2.5 μm, 3 mm X 50 mm are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Suitable for proteomic, peptide mapping and synthetic peptide applications.
  • Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c.
Catalog No. 50-152-8605 Supplier Waters Corp Supplier No. 186008982
The XBridge Peptide BEH C18 XP Column, 130Å, 2.5 μm, 3 mm X 50 mm are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Suitable for proteomic, peptide mapping and synthetic peptide applications.
  • Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c.