Learn More
Waters Corp XBridge BEH C18 XP Column, 130Å, 2.5 µm, 4.6 mm X 150 mm, 1/pkg
Catalog No. 501528610 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 130A C18 columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.
Catalog No. 50-152-8610 Supplier Waters Corp Supplier No. 186008987
The XBridge Peptide BEH C18 130A C18 columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.