Learn More
Waters Corp Waters XBridge Column, Peptide BEH C18 XP, 130Å, 2.5 μm
Supplier:  Waters Corp 186009003
Encompass_Preferred
This XBridge Peptide BEH C18 XP Column Method Validation Kit consists of three 3 mm x 150 mm columns. It features a 2.5 μm particle size and 130 Å pore size, designed for high-resolution peptide separations. The kit is optimized and QC tested for peptide separations, offering a wide usable pH range with superior low pH stability.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.
  • Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c before being QC accepted as a peptide separation column.
  • Method Validation Kit includes three columns from different synthesized batches.
Catalog No. 50-152-8614