Learn More
Waters Corp XBridge Column, C18, 2.5 μm, 130 Å, 4.6 mm X 150 mm
Catalog No. 501528615 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 XP Column, 130Å, 2.5 μm, 4.6 mm X 150 mm MVK columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.
  • Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c before being QC accepted as a Peptide separation column.
  • This Method Validation Kit includes three columns from different synthesized batches.
Catalog No. 50-152-8615 Supplier Waters Corp Supplier No. 186009004
The XBridge Peptide BEH C18 XP Column, 130Å, 2.5 μm, 4.6 mm X 150 mm MVK columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomic, peptide mapping, and synthetic peptide applications.
  • Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c before being QC accepted as a Peptide separation column.
  • This Method Validation Kit includes three columns from different synthesized batches.