Learn More
Waters Corp WATERS CORPORATION XBridge BEH C18 Column, 2.5 μm, 2.1 mm, 75 mm, 130 Å
Catalog No. 50179469 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 column is designed for HPLC method development, offering a broad mobile phase pH range and superior stability. Its advanced particle technology ensures exceptional separation performance and robustness, facilitating seamless transitions between HPLC and UPLC applications.
  • Operates across a mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Offers wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns for exceptional separation performance and throughput.
  • Enables seamless transition toward UPLC adoption.
  • Robust under high pH conditions due to BEH particle synthesis and improved bonding.
Catalog No. 50-179-469 Supplier Waters Corp Supplier No. 186006030
The XBridge BEH C18 column is designed for HPLC method development, offering a broad mobile phase pH range and superior stability. Its advanced particle technology ensures exceptional separation performance and robustness, facilitating seamless transitions between HPLC and UPLC applications.
  • Operates across a mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Offers wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns for exceptional separation performance and throughput.
  • Enables seamless transition toward UPLC adoption.
  • Robust under high pH conditions due to BEH particle synthesis and improved bonding.