Learn More
Waters Corp Waters Column, C18, 2.5 μm, 3 × 30 mm
Catalog No. 50179209 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column offers a universal solution for HPLC method development and separations, featuring a trifunctionally bonded C18 ligand. It provides a wide usable pH range (1-12), superior low pH stability, and ultra-low column bleed, delivering exceptional performance and throughput for HPLC assays.
  • Trifunctionally bonded C18 ligand provides a versatile solution for most HPLC separations.
  • Usable over an entire range of mobile phase pH (1-12).
  • Offers superior low pH stability and ultra-low column bleed.
  • 2.5 μm XP columns deliver exceptional separation performance, robustness, and high throughput.
  • Enables a smooth transition path towards UPLC adoption.
Catalog No. 50-179-209 Supplier Waters Corp Supplier No. 176002550
This column offers a universal solution for HPLC method development and separations, featuring a trifunctionally bonded C18 ligand. It provides a wide usable pH range (1-12), superior low pH stability, and ultra-low column bleed, delivering exceptional performance and throughput for HPLC assays.
  • Trifunctionally bonded C18 ligand provides a versatile solution for most HPLC separations.
  • Usable over an entire range of mobile phase pH (1-12).
  • Offers superior low pH stability and ultra-low column bleed.
  • 2.5 μm XP columns deliver exceptional separation performance, robustness, and high throughput.
  • Enables a smooth transition path towards UPLC adoption.