Learn More
Waters Corp Waters Corporation XBridge BEH C18 Column, 2.5 μm, 3 mm x 30 mm
Supplier:  Waters Corp 186006032
Encompass_Preferred
The XBridge BEH C18 sorbent is designed for HPLC method development, usable across a mobile phase pH range of 1 to 12. Its C18 ligand provides a universal column for most HPLC separations with wide pH range, superior low pH stability, and ultra–low column bleed. When packed in ultra–low dispersion hardware, the 2.5 μm eXtended Performance columns deliver exceptional separation, robustness, and throughput for HPLC assays, also enabling seamless transition to UPLC.
  • C18 chemistry.
  • Reversed phase separation mode.
  • pH range 1 to 12.
  • Maximum pressure 18000 psi (1240 Bar).
  • Particle size 2.5 μm.
  • Universal column for most HPLC separations.
  • Superior low pH stability.
  • Ultra–low column bleed.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables seamless transition towards UPLC adoption.
Catalog No. 50-179-471