Learn More
Waters Corp Waters Column, BEH C18, 2.5 μm, 3 mm X 50 mm
Catalog No. 50179472 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The column is designed for HPLC method development, offering a broad mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand provides superior low pH stability and ultra-low column bleed, enabling robust and high-throughput separations with a seamless transition to UPLC.
  • Provides a wide usable pH range of 1 to 12 with superior low pH stability and ultra-low column bleed.
  • Delivers exceptional separation performance, robustness, and throughput for HPLC and UPLC assays.
  • Maximizes productivity with improved pH stability, enhanced reliability, and particle efficiency.
  • Compatible with all HPLC, UHPLC, and UPLC technology platforms.
  • Features Ethylene Bridged Hybrid (BEH) technology for extreme performance and long lifetime.
  • Supports a simple reversed-phase method development using two mobile-phase pHs and two organic solvents.
Catalog No. 50-179-472 Supplier Waters Corp Supplier No. 186006033
The column is designed for HPLC method development, offering a broad mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand provides superior low pH stability and ultra-low column bleed, enabling robust and high-throughput separations with a seamless transition to UPLC.
  • Provides a wide usable pH range of 1 to 12 with superior low pH stability and ultra-low column bleed.
  • Delivers exceptional separation performance, robustness, and throughput for HPLC and UPLC assays.
  • Maximizes productivity with improved pH stability, enhanced reliability, and particle efficiency.
  • Compatible with all HPLC, UHPLC, and UPLC technology platforms.
  • Features Ethylene Bridged Hybrid (BEH) technology for extreme performance and long lifetime.
  • Supports a simple reversed-phase method development using two mobile-phase pHs and two organic solvents.