Learn More
Waters Corp WATERS CORPORATION Column, BEH C18, 2.5 μm, 2.1 mm x 75 mm, 130 Å
Catalog No. 50179207 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 column offers robust and versatile performance for HPLC method development. With a trifunctionally bonded C18 ligand and eXtended Performance (XP) design, it ensures superior separation and seamless transition capabilities between HPLC and UPLC.
  • Wide mobile phase pH range (1 to 12) for diverse HPLC method development.
  • Trifunctionally bonded C18 ligand provides universal HPLC separations.
  • Superior low pH stability and ultra-low column bleed for robust performance.
  • Ultra-low dispersion hardware and 2.5 μm XP particles for exceptional separation and throughput.
  • Manufactured under strict cGMP and ISO 9002 conditions for high quality and consistent results.
Catalog No. 50-179-207 Supplier Waters Corp Supplier No. 176002548
The XBridge BEH C18 column offers robust and versatile performance for HPLC method development. With a trifunctionally bonded C18 ligand and eXtended Performance (XP) design, it ensures superior separation and seamless transition capabilities between HPLC and UPLC.
  • Wide mobile phase pH range (1 to 12) for diverse HPLC method development.
  • Trifunctionally bonded C18 ligand provides universal HPLC separations.
  • Superior low pH stability and ultra-low column bleed for robust performance.
  • Ultra-low dispersion hardware and 2.5 μm XP particles for exceptional separation and throughput.
  • Manufactured under strict cGMP and ISO 9002 conditions for high quality and consistent results.