Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge BEH C18 Column, 130Å, 2.5 μm, 4.6 mm × 75 mm

Supplier:  Waters Corp 186006038

Encompass_Preferred

Ideal for HPLC method development, this C18 sorbent offers a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand provides broad applicability with superior low pH stability and ultra-low column bleed. The 2.5 μm eXtended Performance [XP] columns ensure exceptional separation performance, robustness, and throughput for HPLC assays, facilitating UPLC transition.

  • C18 chemistry, reversed phase separation
  • Hybrid, spherical particle substrate
  • pH range: 1 to 12
  • Max pressure: 10000 psi (690 Bar)
  • Endcapped, low silanol activity
  • Particle size: 2.5 μm; Pore size: 130 Å
  • ID: 4.6 mm; Length: 75 mm
  • UPLC/UHPLC compatible
  • Superior low pH stability, ultra-low column bleed
  • Exceptional separation performance and robustness

Catalog No. 50-201-516


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.