Learn More
Waters Corp Waters Corporation XBridge BEH C18 Column, 2.5 μm, 4.6 mm x 50 mm, 130 Å
Supplier:  Waters Corp 186006037
Encompass_Preferred
The XBridge BEH C18 column is for HPLC/UHPLC method development. It offers a wide mobile phase pH range (1-12) with superior low pH stability and ultra-low column bleed. Utilizing 2.5 μm XP technology, these columns deliver exceptional separation performance and robustness, facilitating UPLC transition.
  • For HPLC/UHPLC method development.
  • Wide mobile phase pH range (1–12).
  • Superior low pH stability and ultra-low column bleed.
  • Exceptional separation performance and robustness.
  • Compatible with HPLC, UHPLC, and UPLC.
  • Manufactured in cGMP ISO 9001 facility for reproducibility.
  • Individually tested; includes Performance Chromatogram.
Catalog No. 50-201-515