Learn More
Waters Corp Waters Corporation Column, C18, 2.5 μm, 3 mm, 100 mm
Supplier:  Waters Corp 176002553
Encompass_Preferred
The XBridge BEH C18 XP Column is engineered for high-performance liquid chromatography (HPLC) method development, offering robust separation performance across a wide pH range.
  • Supports a mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand provides universal applicability and low pH stability.
  • 2.5 μm eXtended Performance (XP) particles ensure exceptional separation performance.
  • Enables seamless transition to UPLC systems.
  • Proprietary bonding and end-capping technology reduces retention loss at low pH.
  • Ethylene bridges within the silica matrix resist particle dissolution.
  • Manufactured in cGMP, ISO9001, and ISO 13485 certified facilities for outstanding reproducibility.
  • BEH technology offers greater hydrolytic stability.
Catalog No. 50-179-212