Learn More
Waters Corp Waters Corporation Column, C18, 2.5 μm, 3 × 50 mm, 3/pk
Catalog No. 50179578 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit offers three batches of XBridge BEH C18 columns, providing superior separation performance and resilience for HPLC and UPLC applications. These columns withstand pressures up to 18000 psi and feature a tri-functionally bound C18 sorbent, ensuring stability across a broad pH range and extended column life.
  • Superior separation performance and throughput.
  • Seamless HPLC-to-UPLC and UPLC-to-HPLC transfers.
  • Tri-functionally bound C18 sorbent.
  • Wide mobile pH range (1–12) with excellent low pH stability.
  • Reduced column bleed; prevents retention loss and peak shape degradation.
  • Exceptionally long column lifetime.
  • Optimized particle efficiency for excellent peak shape.
  • Manufactured under cGMP and ISO 9002 standards.
Catalog No. 50-179-578 Supplier Waters Corp Supplier No. 186006199
This method validation kit offers three batches of XBridge BEH C18 columns, providing superior separation performance and resilience for HPLC and UPLC applications. These columns withstand pressures up to 18000 psi and feature a tri-functionally bound C18 sorbent, ensuring stability across a broad pH range and extended column life.
  • Superior separation performance and throughput.
  • Seamless HPLC-to-UPLC and UPLC-to-HPLC transfers.
  • Tri-functionally bound C18 sorbent.
  • Wide mobile pH range (1–12) with excellent low pH stability.
  • Reduced column bleed; prevents retention loss and peak shape degradation.
  • Exceptionally long column lifetime.
  • Optimized particle efficiency for excellent peak shape.
  • Manufactured under cGMP and ISO 9002 standards.