Learn More
Waters Corp Waters Corporation XBridge BEH C18 Column, 130Å, 2.5 μm, 2.1 mm X 30 mm
Catalog No. 50179467 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 XP Column is a versatile tool for HPLC method development, offering a wide mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand ensures superior low pH stability and ultra-low column bleed, making it suitable for most HPLC separations.
  • Exceptional separation performance and robustness with ultra-low dispersion hardware.
  • Seamless transition path towards UPLC adoption.
  • Unmatched column lifetime across a wide range of mobile phase pH and temperatures.
  • Improved pH stability for extended column lifetimes and enhanced reliability.
  • Delivers excellent peak shape and capacity by maximizing particle efficiency.
  • Withstands pressures up to 18,000 psi (1,240 bar) without performance deterioration.
Catalog No. 50-179-467 Supplier Waters Corp Supplier No. 186006028
The XBridge BEH C18 XP Column is a versatile tool for HPLC method development, offering a wide mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand ensures superior low pH stability and ultra-low column bleed, making it suitable for most HPLC separations.
  • Exceptional separation performance and robustness with ultra-low dispersion hardware.
  • Seamless transition path towards UPLC adoption.
  • Unmatched column lifetime across a wide range of mobile phase pH and temperatures.
  • Improved pH stability for extended column lifetimes and enhanced reliability.
  • Delivers excellent peak shape and capacity by maximizing particle efficiency.
  • Withstands pressures up to 18,000 psi (1,240 bar) without performance deterioration.