Learn More
Waters Corp Waters Method Validation Kit, BEH C18, 130Å, 2.5 μm, 2.1 mm × 50 mm
Catalog No. 50179576 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns, each packed with a different batch of sorbent. It is designed for reversed-phase separation with C18 chemistry. Key documents, such as Certificates of Analysis, are available online for easy access and reduced waste.
  • Designed for reversed-phase separation with C18 chemistry.
  • Features a hybrid particle substrate with spherical shape and low silanol activity.
  • Supports a broad pH range from 1 to 12.
  • Withstands a maximum pressure of 18000 psi (1240 Bar).
  • Columns are endcapped.
  • Utilizes 2.5 μm particle size and 130 Å pore size.
  • Compatible with UPLC and UHPLC systems.
  • Enhances separation performance and enables rapid method optimization.
  • Provides superior loading capacity for complex mixtures.
Catalog No. 50-179-576 Supplier Waters Corp Supplier No. 186006197
This method validation kit includes three columns, each packed with a different batch of sorbent. It is designed for reversed-phase separation with C18 chemistry. Key documents, such as Certificates of Analysis, are available online for easy access and reduced waste.
  • Designed for reversed-phase separation with C18 chemistry.
  • Features a hybrid particle substrate with spherical shape and low silanol activity.
  • Supports a broad pH range from 1 to 12.
  • Withstands a maximum pressure of 18000 psi (1240 Bar).
  • Columns are endcapped.
  • Utilizes 2.5 μm particle size and 130 Å pore size.
  • Compatible with UPLC and UHPLC systems.
  • Enhances separation performance and enables rapid method optimization.
  • Provides superior loading capacity for complex mixtures.