Learn More
Waters Corp XBridge BEH C18 XP Column, 130Å, 2.5 µm, 4.6 mm X 30 mm, 1/pk
Catalog No. 50201514 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-201-514 Supplier Waters Corp Supplier No. 186006036
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.