Learn More
Waters Corp Waters Column, C18, 130Å, 2.5 μm, 2.1 mm x 50 mm, 3/pk
Catalog No. 50179206 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C18 sorbent is suitable for HPLC method development and can be used across a mobile phase pH range of 1 to 12. The trifunctionally bonded C18 ligand offers a universal column for most HPLC separations, providing a wide usable pH range, superior low pH stability, and ultra-low column bleed. When packed in ultra-low dispersion hardware, the 2.5 μm columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, while facilitating a seamless transition to UPLC adoption.
  • Widest usable pH range (1 to 12)
  • Superior low pH stability and ultra-low column bleed
  • Exceptional separation performance, robustness, and throughput for HPLC assays
  • Seamless transition path toward UPLC adoption
  • Excellent peak shape and capacity
  • Extraordinary column lifetime
Catalog No. 50-179-206 Supplier Waters Corp Supplier No. 176002547
Add to Cart
Add to Cart
This C18 sorbent is suitable for HPLC method development and can be used across a mobile phase pH range of 1 to 12. The trifunctionally bonded C18 ligand offers a universal column for most HPLC separations, providing a wide usable pH range, superior low pH stability, and ultra-low column bleed. When packed in ultra-low dispersion hardware, the 2.5 μm columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, while facilitating a seamless transition to UPLC adoption.
  • Widest usable pH range (1 to 12)
  • Superior low pH stability and ultra-low column bleed
  • Exceptional separation performance, robustness, and throughput for HPLC assays
  • Seamless transition path toward UPLC adoption
  • Excellent peak shape and capacity
  • Extraordinary column lifetime