Learn More
Waters Corp Waters Column, C8, 2.5 μm, 2.1 mm, 150 mm
Supplier: Waters Corp 186006712
The XBridge BEH C8 sorbent, less retentive than C18, enables faster method development, improved peak shape, and better chromatographic performance. This trifunctionally bonded sorbent offers a broad usable pH range (1 to 12), excellent low pH stability, and minimal column bleed. The 2.5 μm XP columns deliver exceptional separation, robustness, and throughput for HPLC, allowing seamless transition to UPLC.
- Maximizes productivity for quality-control and advanced LC-MS assays.
- Delivers consistent, high-quality results; manufactured under cGMP and ISO 9001 standards.
- Packing materials ensure excellent peak shape, high efficiency, and stability.
- Chemistry: C8, Reversed Phase, Hybrid particle substrate.
- Particle size: 2.5 μm; Inner diameter: 2.1 mm; Length: 150 mm.
- Pore size: 130 Å; Max pressure: 18000 psi (1240 Bar).
- Endcapped with low silanol activity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.