Learn More
Waters Corp XBridge Method Validation Kit, C8, 2.5 μm, 2.1 mm × 150 mm, 3 pk
Catalog No. 50466462 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three XBridge BEH C8 columns, each packed with a different batch of sorbent. The XBridge BEH C8 sorbent offers less retention compared to C18, allowing for faster method development. It provides superior stability at low pH and ultra-low column bleed, and is designed to deliver exceptional separation performance for HPLC assays with a seamless transition to UPLC.
  • Less retentive C8 sorbent allows for faster method development.
  • Trifunctionally bonded sorbent provides a wide usable pH range of 1 to 12.
  • Superior stability at low pH for robust analytical methods.
  • Ultra-low column bleed ensures high-quality data.
  • Packed in ultra-low dispersion hardware for exceptional separation performance.
  • Compatible with UPLC and UHPLC systems.
  • Digital certificates of analysis are available online, eliminating paper documentation.
Catalog No. 50-466-462 Supplier Waters Corp Supplier No. 186006760
This method validation kit includes three XBridge BEH C8 columns, each packed with a different batch of sorbent. The XBridge BEH C8 sorbent offers less retention compared to C18, allowing for faster method development. It provides superior stability at low pH and ultra-low column bleed, and is designed to deliver exceptional separation performance for HPLC assays with a seamless transition to UPLC.
  • Less retentive C8 sorbent allows for faster method development.
  • Trifunctionally bonded sorbent provides a wide usable pH range of 1 to 12.
  • Superior stability at low pH for robust analytical methods.
  • Ultra-low column bleed ensures high-quality data.
  • Packed in ultra-low dispersion hardware for exceptional separation performance.
  • Compatible with UPLC and UHPLC systems.
  • Digital certificates of analysis are available online, eliminating paper documentation.